Merck
A structure-based designed small molecule depletes hRpn13Pru and a select group of KEN box proteins.
Nature communications (2024-03-21)
Xiuxiu Lu, Monika Chandravanshi, Venkata R Sabbasani, Snehal Gaikwad, V Keith Hughitt, Nana Gyabaah-Kessie, Bradley T Scroggins, Sudipto Das, Wazo Myint, Michelle E Clapp, Charles D Schwieters, Marzena A Dyba, Derek L Bolhuis, Janusz W Koscielniak, Thorkell Andresson, Michael J Emanuele, Nicholas G Brown, Hiroshi Matsuo, Raj Chari, Deborah E Citrin, Beverly A Mock, Rolf E Swenson, Kylie J Walters
ABSTRAKT
Proteasome subunit hRpn13 is partially proteolyzed in certain cancer cell types to generate hRpn13Pru by degradation of its UCHL5/Uch37-binding DEUBAD domain and retention of an intact proteasome- and ubiquitin-binding Pru domain. By using structure-guided virtual screening, we identify an hRpn13 binder (XL44) and solve its structure ligated to hRpn13 Pru by integrated X-ray crystallography and NMR to reveal its targeting mechanism. Surprisingly, hRpn13Pru is depleted in myeloma cells following treatment with XL44. TMT-MS experiments reveal a select group of off-targets, including PCNA clamp-associated factor PCLAF and ribonucleoside-diphosphate reductase subunit M2 (RRM2), that are similarly depleted by XL44 treatment. XL44 induces hRpn13-dependent apoptosis and also restricts cell viability by a PCLAF-dependent mechanism. A KEN box, but not ubiquitination, is required for XL44-induced depletion of PCLAF. Here, we show that XL44 induces ubiquitin-dependent loss of hRpn13Pru and ubiquitin-independent loss of select KEN box containing proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution