Merck
Small molecule targeting of transcription-replication conflict for selective chemotherapy.
Cell chemical biology (2023-08-03)
Long Gu, Min Li, Caroline M Li, Pouya Haratipour, Robert Lingeman, Jennifer Jossart, Margarita Gutova, Linda Flores, Caitlyn Hyde, Nikola Kenjić, Haiqing Li, Vincent Chung, Hongzhi Li, Brett Lomenick, Daniel D Von Hoff, Timothy W Synold, Karen S Aboody, Yilun Liu, David Horne, Robert J Hickey, J Jefferson P Perry, Linda H Malkas
ABSTRAKT
Targeting transcription replication conflicts, a major source of endogenous DNA double-stranded breaks and genomic instability could have important anticancer therapeutic implications. Proliferating cell nuclear antigen (PCNA) is critical to DNA replication and repair processes. Through a rational drug design approach, we identified a small molecule PCNA inhibitor, AOH1996, which selectively kills cancer cells. AOH1996 enhances the interaction between PCNA and the largest subunit of RNA polymerase II, RPB1, and dissociates PCNA from actively transcribed chromatin regions, while inducing DNA double-stranded breaks in a transcription-dependent manner. Attenuation of RPB1 interaction with PCNA, by a point mutation in RPB1's PCNA-binding region, confers resistance to AOH1996. Orally administrable and metabolically stable, AOH1996 suppresses tumor growth as a monotherapy or as a combination treatment but causes no discernable side effects. Inhibitors of transcription replication conflict resolution may provide a new and unique therapeutic avenue for exploiting this cancer-selective vulnerability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-DNA-RNA Hybrid Antibody, clone S9.6, clone S9.6, from mouse
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution