Merck
Microglial cGAS drives neuroinflammation in the MPTP mouse models of Parkinson's disease.
CNS neuroscience & therapeutics (2023-03-14)
Chunmei Ma, Ying Liu, Sheng Li, Chanyuan Ma, Jiajia Huang, Shuang Wen, Shuo Yang, Bingwei Wang
ABSTRAKT
Neuroinflammation has been widely accepted as a cause of the degenerative process. Increasing interest has been devoted to developing intervening therapeutics for preventing neuroinflammation in Parkinson's disease (PD). It is well known that virus infections, including DNA viruses, are associated with an increased risk of PD. In addition, damaged or dying dopaminergic neurons can release dsDNA during PD progression. However, the role of cGAS, a cytosolic dsDNA sensor, in PD progression remains unclear. Adult male wild-type mice and age-matched male cGAS knockout (cGas-/- ) mice were treated with MPTP to induce neurotoxic PD model, and then behavioral tests, immunohistochemistry, and ELISA were conducted to compare disease phenotype. Chimeric mice were reconstituted to explore the effects of cGAS deficiency in peripheral immune cells or CNS resident cells on MPTP-induced toxicity. RNA sequencing was used to dissect the mechanistic role of microglial cGAS in MPTP-induced toxicity. cGAS inhibitor administration was conducted to study whether GAS may serve as a therapeutic target. We observed that the cGAS-STING pathway was activated during neuroinflammation in MPTP mouse models of PD. cGAS deficiency in microglia, but not peripheral immune cells, controlled neuroinflammation and neurotoxicity induced by MPTP. Mechanistically, microglial cGAS ablation alleviated the neuronal dysfunction and inflammatory response in astrocytes and microglia by inhibiting antiviral inflammatory signaling. Additionally, the administration of cGAS inhibitors conferred the mice neuroprotection during MPTP exposure. Collectively, these findings demonstrate microglial cGAS promote neuroinflammation and neurodegeneration during the progression of MPTP-induced PD mouse models and suggest cGAS may serve as a therapeutic target for PD patients. Although we demonstrated that cGAS promotes the progression of MPTP-induced PD, this study has limitations. We identified that cGAS in microglia accelerate disease progression of PD by using bone marrow chimeric experiments and analyzing cGAS expression in CNS cells, but evidence would be more straightforward if conditional knockout mice were used. This study contributed to the knowledge of the role of the cGAS pathway in PD pathogenesis; nevertheless, trying more PD animal models in the future will help us to understand the disease progression deeper and explore possible treatments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture