Merck
Comparison of Natural Killer Cells Differentiated from Various Pluripotent Stem Cells.
International journal of molecular sciences (2024-08-10)
Jongsuk Han, Hyeongbin Son, Daun Jung, Ki-Yeon Kim, Chaeyeon Jin, Hyeonwook Hwang, Soon-Suk Kang, Shoukhrat Mitalipov, Hee-Jung An, Yeonmi Lee, Eunju Kang
ABSTRAKT
Allogeneic natural killer (NK) cell therapy has been effective in treating cancer. Many studies have tested NK cell therapy using human pluripotent stem cells (hPSCs). However, the impacts of the origin of PSC-NK cells on competence are unclear. In this study, several types of hPSCs, including human-induced PSCs (hiPSCs) generated from CD34+, CD3-CD56+, and CD56- cells in umbilical cord blood (UCB), three lines of human embryonic stem cells (hESCs, ES-1. ES-2 and ES-3) and MHC I knockout (B2M-KO)-ESCs were used to differentiate into NK cells and their capacities were analyzed. All PSC types could differentiate into NK cells. Among the iPSC-derived NK cells (iPSC-NKs) and ESC-derived NK cells (ES-NKs), 34+ iPSCs and ES-3 had a higher growth rate and cytotoxicity, respectively, ES-3 also showed better efficacy than 34+ iPSCs. B2M-KO was similar to the wild type. These results suggest that the screening for differentiation of PSCs into NK cells prior to selecting the PSC lines for the development of NK cell immunotherapy is an essential process for universal allotransplantation, including the chimeric antigen receptor (CAR).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture