Merck
Control of cognition and adaptive behavior by the GLP/G9a epigenetic suppressor complex.
Neuron (2009-12-17)
Anne Schaefer, Srihari C Sampath, Adam Intrator, Alice Min, Tracy S Gertler, D James Surmeier, Alexander Tarakhovsky, Paul Greengard
ABSTRAKT
The genetic basis of cognition and behavioral adaptation to the environment remains poorly understood. Here we demonstrate that the histone methyltransferase complex GLP/G9a controls cognition and adaptive responses in a region-specific fashion in the adult brain. Using conditional mutagenesis in mice, we show that postnatal, neuron-specific deficiency of GLP/G9a leads to derepression of numerous nonneuronal and neuron progenitor genes in adult neurons. This transcriptional alteration is associated with complex behavioral abnormalities, including defects in learning, motivation, and environmental adaptation. The behavioral changes triggered by GLP/G9a deficiency are similar to key symptoms of the human 9q34 mental retardation syndrome that is associated with structural alterations of the GLP/EHMT1 gene. The likely causal role of GLP/G9a in mental retardation in mice and humans suggests a key role for the GLP/G9a-controlled histone H3K9 dimethylation in regulation of brain function through maintenance of the transcriptional homeostasis in adult neurons.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride, for Membrane Applications