Merck
Phenotypic switching as a non-genetic mechanism of resistance predicts antibody therapy regimens.
iScience (2024-03-28)
Jiawei Zhou, Can Liu, Yu Tang, Zhongbo Li, Yanguang Cao
ABSTRAKT
Despite the specificity and effectiveness of antibody therapy, resistance to treatment remains a major barrier for their broad clinical applications. While genetic mutations are known to be critical, the impact of non-genetic mechanisms, such as epigenetic changes and phenotypic adaptations, on resistance to antibody-dependent cellular cytotoxicity (ADCC) is not fully understood. Our study investigated the non-genetic resistance mechanisms that colorectal cancer cells develop against cetuximab and the resulting ADCC pressure. Resistance clones exhibited decreased EGFR/HER2 expressions, enriched interferon-related pathways, and lower NK cell activation. Interestingly, these resistance clones regained sensitivity upon the withdrawal of therapeutic pressure, implying phenotypic plasticity and reversibility. To counter resistance, we developed a mathematical model recapitulating the phenotypic switching dynamics. The model predicted that intermittent dosing strategy outperforms continuous regimen in delaying treatment resistance. Our findings have implications for improving efficacy and circumventing resistance to targeted antibody therapies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture