Merck
Age-specific nasal epithelial responses to SARS-CoV-2 infection.
Nature microbiology (2024-04-16)
Maximillian N J Woodall, Ana-Maria Cujba, Kaylee B Worlock, Katie-Marie Case, Tereza Masonou, Masahiro Yoshida, Krzysztof Polanski, Ni Huang, Rik G H Lindeboom, Lira Mamanova, Liam Bolt, Laura Richardson, Batuhan Cakir, Samuel Ellis, Machaela Palor, Thomas Burgoyne, Andreia Pinto, Dale Moulding, Timothy D McHugh, Aarash Saleh, Eliz Kilich, Puja Mehta, Chris O'Callaghan, Jie Zhou, Wendy Barclay, Paolo De Coppi, Colin R Butler, Mario Cortina-Borja, Heloise Vinette, Sunando Roy, Judith Breuer, Rachel C Chambers, Wendy E Heywood, Kevin Mills, Robert E Hynds, Sarah A Teichmann, Kerstin B Meyer, Marko Z Nikolić, Claire M Smith
ABSTRAKT
Children infected with SARS-CoV-2 rarely progress to respiratory failure. However, the risk of mortality in infected people over 85 years of age remains high. Here we investigate differences in the cellular landscape and function of paediatric (<12 years), adult (30-50 years) and older adult (>70 years) ex vivo cultured nasal epithelial cells in response to infection with SARS-CoV-2. We show that cell tropism of SARS-CoV-2, and expression of ACE2 and TMPRSS2 in nasal epithelial cell subtypes, differ between age groups. While ciliated cells are viral replication centres across all age groups, a distinct goblet inflammatory subtype emerges in infected paediatric cultures and shows high expression of interferon-stimulated genes and incomplete viral replication. In contrast, older adult cultures infected with SARS-CoV-2 show a proportional increase in basaloid-like cells, which facilitate viral spread and are associated with altered epithelial repair pathways. We confirm age-specific induction of these cell types by integrating data from in vivo COVID-19 studies and validate that our in vitro model recapitulates early epithelial responses to SARS-CoV-2 infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Mucin MUC5AC Antibody, clone CLH2, clone CLH2, Chemicon®, from mouse