Merck
  • Quantifying nanoscopic alterations associated with mitochondrial dysfunction using three-dimensional single-molecule localization microscopy.
Quantifying nanoscopic alterations associated with mitochondrial dysfunction using three-dimensional single-molecule localization microscopy.
Biomedical optics express (2024-03-18)
Benjamin Brenner, Fengyuanshan Xu, Yang Zhang, Junghun Kweon, Raymond Fang, Nader Sheibani, Sarah X Zhang, Cheng Sun, Hao F Zhang
ABSTRAKT
Mitochondrial morphology provides unique insights into their integrity and function. Among fluorescence microscopy techniques, 3D super-resolution microscopy uniquely enables the analysis of mitochondrial morphological features individually. However, there is a lack of tools to extract morphological parameters from super-resolution images of mitochondria. We report a quantitative method to extract mitochondrial morphological metrics, including volume, aspect ratio, and local protein density, from 3D single-molecule localization microscopy images, with single-mitochondrion sensitivity. We validated our approach using simulated ground-truth SMLM images of mitochondria. We further tested our morphological analysis on mitochondria that have been altered functionally and morphologically in controlled manners. This work sets the stage to quantitatively analyze mitochondrial morphological alterations associated with disease progression on an individual basis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-TOMM20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution