Merck
  • Proteomic Identification of Small Extracellular Vesicle Proteins LAMB1 and Histone H4 for Prostate Cancer Diagnosis and Risk Stratification.
Proteomic Identification of Small Extracellular Vesicle Proteins LAMB1 and Histone H4 for Prostate Cancer Diagnosis and Risk Stratification.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2024-04-09)
Bairen Pang, Qi Wang, Haotian Chen, Zhihan Liu, Meng Han, Jie Gong, Liang Yue, Xuan Ding, Suying Wang, Zejun Yan, Yingzhi Chen, David Malouf, Joseph Bucci, Tiannan Guo, Cheng Zhou, Junhui Jiang, Yong Li
ABSTRAKT
Diagnosis and stratification of prostate cancer (PCa) patients using the prostate-specific antigen (PSA) test is challenging. Extracellular vesicles (EVs), as a new star of liquid biopsy, has attracted interest to complement inaccurate PSA screening and invasiveness of tissue biopsy. In this study, a panel of potential small EV (sEV) protein biomarkers is identified from PCa cell lines using label-free LC-MS/MS proteomics. These biomarkers underwent further validation with plasma and urine samples from different PCa stages through parallel reaction monitoring-based targeted proteomics, western blotting, and ELISA. Additionally, a tissue microarray containing cancerous and noncancerous tissues is screened to provide additional evidence of selected sEV proteins associated with cancer origin. Results indicate that sEV protein LAMB1 is highly expressed in human plasma of metastatic PCa patients compared with localised PCa patients and control subjects, while sEV protein Histone H4 is highly expressed in human urine of high-risk PCa patients compared to low-risk PCa patients and control subjects. These two sEV proteins demonstrate higher specificity and sensitivity than the PSA test and show promise for metastatic PCa diagnosis, progression monitoring, and risk stratification.