Merck
Peroxisome proliferator-activated receptor-gamma expression in the lung tissue of obese rats.
Yonsei medical journal (2011-04-14)
Su Jin Hwang, Jung Ho Kim, Jae Won Shim, Deok Soo Kim, Duk Soo Kim, Hye Lim Jung, Moon Soo Park, Won Young Lee, Se-Yeon Kim, Jung Yeon Shim
ABSTRAKT
Obesity is a risk factor for asthma and type II diabetes. Peroxisome proliferator- activated receptor (PPAR)-γ has been suggested to regulate inflammatory responses in diabetes and asthma. We investigated whether PPAR-α, PPAR-γ, adiponectin receptors (AdipoR1, AdipoR2), leptin, and tumor necrosis factor (TNF)-α are expressed in rat lung tissues and whether the expression differs between obese Otsuka Long-Evans Tokushima Fatty (OLETF) and lean Long Evans Tokushima Otsuka (LETO) rats. Obese and lean rats were given with a high fat diet or a 30% restricted diet for 32 weeks, and their blood glucose levels and weights were monitored. After 32 weeks, mRNA levels of PPAR-α, PPAR-γ, AdipoR1, AdipoR2, leptin, and TNF-α in lung tissues were measured using real time PCR. PPAR-α, PPAR-γ, AdipoR1, AdipoR2, leptin, and TNF-α were expressed in both obese and lean rat lung tissues. Increased serum glucose levels on intraperitoneal glucose tolerance testing and a higher weight gain at 32 weeks were observed in OLETF control rats compared to OLETF diet restricted rats. PPAR-γ expression was markedly elevated in obese control and diet restricted rats compared to lean rats, although PPAR-γ expression in obese rats was not affected by diet restriction. Leptin was highly expressed in OLETF rats compared to LETO rats. TNF-α expression was enhanced in OLETF control rats compared LETO diet restricted rats, and decreased by diet restriction. PPAR-α, AdipoR1, and AdipoR2 expression were not significantly different between obese and lean rats. PPAR-γ was highly expressed in the lung tissues of obese rats and may be a novel treatment target for regulating lung inflammation associated with obesity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sigma-Aldrich
TRI Reagent®
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.