Merck
Reprogramming mechanism dissection and trophoblast replacement application in monkey somatic cell nuclear transfer.
Nature communications (2024-01-17)
Zhaodi Liao, Jixiang Zhang, Shiyu Sun, Yuzhuo Li, Yuting Xu, Chunyang Li, Jing Cao, Yanhong Nie, Zhuoyue Niu, Jingwen Liu, Falong Lu, Zhen Liu, Qiang Sun
ABSTRAKT
Somatic cell nuclear transfer (SCNT) successfully clones cynomolgus monkeys, but the efficiency remains low due to a limited understanding of the reprogramming mechanism. Notably, no rhesus monkey has been cloned through SCNT so far. Our study conducts a comparative analysis of multi-omics datasets, comparing embryos resulting from intracytoplasmic sperm injection (ICSI) with those from SCNT. Our findings reveal a widespread decrease in DNA methylation and the loss of imprinting in maternally imprinted genes within SCNT monkey blastocysts. This loss of imprinting persists in SCNT embryos cultured in-vitro until E17 and in full-term SCNT placentas. Additionally, histological examination of SCNT placentas shows noticeable hyperplasia and calcification. To address these defects, we develop a trophoblast replacement method, ultimately leading to the successful cloning of a healthy male rhesus monkey. These discoveries provide valuable insights into the reprogramming mechanism of monkey SCNT and introduce a promising strategy for primate cloning.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.