Merck
Deep brain stimulation alleviates tics in Tourette syndrome via striatal dopamine transmission.
Brain : a journal of neurology (2023-05-04)
Aaron E Rusheen, Juan Rojas-Cabrera, Abhinav Goyal, Hojin Shin, Jason Yuen, Dong-Pyo Jang, Keven E Bennet, Charles D Blaha, Kendall H Lee, Yoonbae Oh
ABSTRAKT
Tourette syndrome is a childhood-onset neuropsychiatric disorder characterized by intrusive motor and vocal tics that can lead to self-injury and deleterious mental health complications. While dysfunction in striatal dopamine neurotransmission has been proposed to underlie tic behaviour, evidence is scarce and inconclusive. Deep brain stimulation (DBS) of the thalamic centromedian parafascicular complex (CMPf), an approved surgical interventive treatment for medical refractory Tourette syndrome, may reduce tics by affecting striatal dopamine release. Here, we use electrophysiology, electrochemistry, optogenetics, pharmacological treatments and behavioural measurements to mechanistically examine how thalamic DBS modulates synaptic and tonic dopamine activity in the dorsomedial striatum. Previous studies demonstrated focal disruption of GABAergic transmission in the dorsolateral striatum of rats led to repetitive motor tics recapitulating the major symptom of Tourette syndrome. We employed this model under light anaesthesia and found CMPf DBS evoked synaptic dopamine release and elevated tonic dopamine levels via striatal cholinergic interneurons while concomitantly reducing motor tic behaviour. The improvement in tic behaviour was found to be mediated by D2 receptor activation as blocking this receptor prevented the therapeutic response. Our results demonstrate that release of striatal dopamine mediates the therapeutic effects of CMPf DBS and points to striatal dopamine dysfunction as a driver for motor tics in the pathoneurophysiology of Tourette syndrome.