Merck
Deletion of Pax1 scoliosis-associated regulatory elements leads to a female-biased tail abnormality.
Cell reports (2024-03-10)
Aki Ushiki, Rory R Sheng, Yichi Zhang, Jingjing Zhao, Mai Nobuhara, Elizabeth Murray, Xin Ruan, Jonathan J Rios, Carol A Wise, Nadav Ahituv
ABSTRAKT
Adolescent idiopathic scoliosis (AIS), a sideways curvature of the spine, is sexually dimorphic, with increased incidence in females. A genome-wide association study identified a female-specific AIS susceptibility locus near the PAX1 gene. Here, we use mouse enhancer assays, three mouse enhancer knockouts, and subsequent phenotypic analyses to characterize this region. Using mouse enhancer assays, we characterize a sequence, PEC7, which overlaps the AIS-associated variant, and find it to be active in the tail tip and intervertebral disc. Removal of PEC7 or Xe1, a known sclerotome enhancer nearby, or deletion of both sequences lead to a kinky tail phenotype only in the Xe1 and combined (Xe1+PEC7) knockouts, with only the latter showing a female sex dimorphic phenotype. Extensive phenotypic characterization of these mouse lines implicates several differentially expressed genes and estrogen signaling in the sex dimorphic bias. In summary, our work functionally characterizes an AIS-associated locus and dissects the mechanism for its sexual dimorphism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
Anty-Digoxigenin-AP, fragmenty Fab, from sheep
Sigma-Aldrich
Tamoxifen, ≥99%