Merck
Cancer-associated fibroblasts produce matrix-bound vesicles that influence endothelial cell function.
Science signaling (2024-03-12)
Alice Santi, Emily J Kay, Lisa J Neilson, Lynn McGarry, Sergio Lilla, Margaret Mullin, Nikki R Paul, Frédéric Fercoq, Grigorios Koulouras, Giovanny Rodriguez Blanco, Dimitris Athineos, Susan Mason, Mark Hughes, Gemma Thomson, Yann Kieffer, Colin Nixon, Karen Blyth, Fatima Mechta-Grigoriou, Leo M Carlin, Sara Zanivan
ABSTRAKT
Intercellular communication between different cell types in solid tumors contributes to tumor growth and metastatic dissemination. The secretome of cancer-associated fibroblasts (CAFs) plays major roles in these processes. Using human mammary CAFs, we showed that CAFs with a myofibroblast phenotype released extracellular vesicles that transferred proteins to endothelial cells (ECs) that affected their interaction with immune cells. Mass spectrometry-based proteomics identified proteins transferred from CAFs to ECs, which included plasma membrane receptors. Using THY1 as an example of a transferred plasma membrane-bound protein, we showed that CAF-derived proteins increased the adhesion of a monocyte cell line to ECs. CAFs produced high amounts of matrix-bound EVs, which were the primary vehicles of protein transfer. Hence, our work paves the way for future studies that investigate how CAF-derived matrix-bound EVs influence tumor pathology by regulating the function of neighboring cancer, stromal, and immune cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid