Merck
Human gut microbiome and metabolite dynamics under simulated microgravity.
Gut microbes (2023-09-26)
Ana Ramos-Nascimento, Lucia Grenga, Sven-Bastiaan Haange, Alexandra Himmelmann, Franca Sabine Arndt, Yen-Tran Ly, Guylaine Miotello, Olivier Pible, Nico Jehmlich, Beatrice Engelmann, Martin von Bergen, Edwin Mulder, Petra Frings-Meuthen, Christine Elisabeth Hellweg, Jens Jordan, Ulrike Rolle-Kampczyk, Jean Armengaud, Ralf Moeller
ABSTRAKT
The Artificial Gravity Bed Rest - European Space Agency (AGBRESA) study was the first joint bed rest study by ESA, DLR, and NASA that examined the effect of simulated weightlessness on the human body and assessed the potential benefits of artificial gravity as a countermeasure in an analog of long-duration spaceflight. In this study, we investigated the impact of simulated microgravity on the gut microbiome of 12 participants during a 60-day head-down tilt bed rest at the :envihab facilities. Over 60 days of simulated microgravity resulted in a mild change in the gut microbiome, with distinct microbial patterns and pathway expression in the feces of the countermeasure group compared to the microgravity simulation-only group. Additionally, we found that the countermeasure protocols selectively increased the abundance of beneficial short-chain fatty acids in the gut, such as acetate, butyrate, and propionate. Some physiological signatures also included the modulation of taxa reported to be either beneficial or opportunistic, indicating a mild adaptation in the microbiome network balance. Our results suggest that monitoring the gut microbial catalog along with pathway clustering and metabolite profiling is an informative synergistic strategy to determine health disturbances and the outcome of countermeasure protocols for future space missions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Butyric acid, ≥99%