Merck
Pentachloropseudilin Treatment Impairs Host Cell Invasion by Trypanosoma cruzi.
Chembiochem : a European journal of chemical biology (2022-07-16)
Mylla Spirandelli da Costa, Bruna Cristina Borges, Isabella Teixeira Marques, Rayane Cristina de Oliveira, Thaise Lara Teixeira, Julia de Gouveia Santos, Claudio Vieira da Silva
ABSTRAKT
Pentachloropseudilin (PClP) is a reversible and allosteric inhibitor of type 1 myosin. Here, we addressed the impact of PClP treatment of Trypanosoma cruzi and mammalian host cell on the parasite migration, cell adhesion and invasion. We observed that PClP was not toxic to either T. cruzi or host cell. Moreover, treatment of T. cruzi with PClP inhabited parasite motility, host cell adhesion and invasion. Treatment of host cell with PClP also impaired parasite invasion probably by decreasing lysosome migration to the entry site of the parasite. Therefore, PClP treatment impaired fundamental processes necessary for a successful T. cruzi infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pentachloropseudilin, ≥98% (HPLC)