Merck
  • Inhibitory effects of the herbal medicine Sho-saiko-to (TJ-9) on cell proliferation and procollagen gene expressions in cultured rat hepatic stellate cells.
Inhibitory effects of the herbal medicine Sho-saiko-to (TJ-9) on cell proliferation and procollagen gene expressions in cultured rat hepatic stellate cells.
Journal of hepatology (1998-11-21)
K Kayano, I Sakaida, K Uchida, K Okita
ABSTRAKT
It is of extreme importance to prevent liver fibrosis and subsequent progression to liver cirrhosis. The aim of our study was to elucidate in vitro whether Sho-saiko-to exerted inhibitory effects on hepatic stellate cells. Hepatic stellate cells were isolated from male Wistar rats. Water-soluble ingredients of Sho-saiko-to were obtained at concentrations of 10, 100, 250, 500 and 1000 microg/ml. Morphological transformation was observed under a phase-contrast microscope. Flow cytometric analysis was performed on day 4 after culture to evaluate the potential to proliferate of the stellate cells by analyzing cell cycles. Northern blot analysis was carried out on day 3 after culture to determine the expressions of type I and type III procollagen mRNAs. (i) Sho-saiko-to 500 and 1000 microg/ml inhibited morphological transformation of the stellate cells to myofibroblast-like cells. (ii) Sho-saiko-to 500 and 1000 microg/ml significantly (p<0.0001) accumulated the cells in the G0/G1 phase (118.8+/-0.7%, 119.2+/-0.5%, respectively as compared with control) and significantly (p<0.0001) decreased cell numbers subsequently in G2/M phase (47.5+/-8.1%, 48.9+/-2.0%, respectively). (iii) Sho-saiko-to 500 and 1000 microg/ml also significantly (p<0.05 or p<0.0001) suppressed procollagen mRNA expression of type I to 51.5+/-6.4%, 34.9+/-3.7%, respectively, and type III to 51.3+/-12.3%, 46.7+/-11.4%, respectively. We have clarified the inhibitory effects of Sho-saiko-to on hepatic stellate cells in vitro. Sho-saiko-to could be a potent inhibitor in the pathogenesis of liver fibrosis.