- Total regio- and diastereocontrol in the aldol reactions of dienolborinates.
Total regio- and diastereocontrol in the aldol reactions of dienolborinates.
Organic letters (2013-03-08)
P Veeraraghavan Ramachandran, Daniel Nicponski, Bomi Kim
PMID23465070
ABSTRAKT
It is reported that appropriate dienolborinates can provide access to both diastereomers of 2-(hydroxymethyl)but-3-enoates through exclusive α-regiocontrol in a non-vinylogous pathway. Contrary to previous reports in which dialkylchloroboranes failed to enolize propanoates, acidity-enhanced but-3-enoates readily undergo enolization, offering unprecedented control over the formation of these valuable synthons. The first example of an aldol reaction in the presence of a phosphine-borane adduct is also reported.