Merck
Activity of etoposide (VP-16) and teniposide (VM-26) in exponential and plateau phase human tumor cell cultures.
Anti-cancer drugs (1992-06-01)
J C Chapuis, P C Keng, D W Siemann
ABSTRAKT
The effects of etoposide (VP-16) and teniposide (VM-26) have been evaluated in human epidermoid carcinoma cells (A431, ME180 and HEp3) grown as exponential and plateau phase cultures. A significant increase in resistance to both these chemotherapeutic agents was observed in unfed plateau compared with exponential phase cells. The large differences in cell killing could not be explained by cell cycle specific toxicities resulting from variations in the cell cycle distributions. Rather the differences in the treatment efficacies probably reflect the 5- to 15-fold increase in the proportion of quiescent cells measured in the plateau phase cultures. These findings suggest that non-proliferating cells in tumors may be preferentially spared in treatments utilizing VP-16 and VM-26.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Linia komórkowa A431, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85090402, human skin, Epithelial