Merck
  • Sensitivity to EBV superinfection and IUdR inducibility of hybrid cells formed between a sensitive and a relatively resistant Burkitt lymphoma cell line.
Sensitivity to EBV superinfection and IUdR inducibility of hybrid cells formed between a sensitive and a relatively resistant Burkitt lymphoma cell line.
International journal of cancer (1973-09-15)
O Nyormoi, G Klein, A Adams, L Dombos
MATERIAŁY
Numer produktu
Marka
Opis produktu
Namalwa, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87060801, human blood, Lymphoblast