- A comparison of the sensitivity of immunofluorescence and Giemsa for staining Chlamydia trachomatis inclusions in cycloheximide-treated McCoy cells.
A comparison of the sensitivity of immunofluorescence and Giemsa for staining Chlamydia trachomatis inclusions in cycloheximide-treated McCoy cells.
Journal of clinical pathology (1980-02-01)
P E Munday, A P Johnson, B J Thomas, D Taylor-Robinson
PMID6988463
ABSTRAKT
In a clinical study of 190 men with non-gonococcal urethritis, Chlamydia trachomatis inclusions were sought in cycloheximide-treated McCoy cells by an indirect immunofluorescent antibody technique. The method was consistently reliable over a period of two years, and the results were obtained within 24 hours of a patient's attendance. The results correlated with those obtained by Giemsa staining in 91.6% of patients, and the new method was at least as sensitive as the established Giemsa-staining method.
MATERIAŁY
Numer produktu
Marka
Opis produktu
McCoy, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90010305, mouse unknown/unspecified, Fibroblast