Merck
Mechanistic study of a low-power bacterial maintenance state using high-throughput electrochemistry.
Cell (2024-10-25)
John A Ciemniecki, Chia-Lun Ho, Richard D Horak, Akihiro Okamoto, Dianne K Newman
ABSTRAKT
Mechanistic studies of life's lower metabolic limits have been limited due to a paucity of tractable experimental systems. Here, we show that redox-cycling of phenazine-1-carboxamide (PCN) by Pseudomonas aeruginosa supports cellular maintenance in the absence of growth with a low mass-specific metabolic rate of 8.7 × 10-4 W (g C)-1 at 25°C. Leveraging a high-throughput electrochemical culturing device, we find that non-growing cells cycling PCN tolerate conventional antibiotics but are susceptible to those that target membrane components. Under these conditions, cells conserve energy via a noncanonical, facilitated fermentation that is dependent on acetate kinase and NADH dehydrogenases. Across PCN concentrations that limit cell survival, the cell-specific metabolic rate is constant, indicating the cells are operating near their bioenergetic limit. This quantitative platform opens the door to further mechanistic investigations of maintenance, a physiological state that underpins microbial survival in nature and disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Phenazine-1-carboxamide, ≥98% (HPLC), powder, antimicrobial