Merck
Trends of fluoroquinolones resistance in Mycoplasma and Ureaplasma urogenital isolates: Systematic review and meta-analysis.
Journal of global antimicrobial resistance (2023-11-29)
Yaping Wu, Nahal Majidzadeh, Ying Li, Matin Zafar Shakourzadeh, Sara Hajilari, Ebrahim Kouhsari, Khalil Azizian
ABSTRAKT
Mycoplasma and Ureaplasma spp. especially M. hominis, U. parvum, and U. urealyticum recognized as an important cause of urogenital infections. Sake of the presence of antibiotic resistance and a continuous rise in resistance, the treatment options are limited, and treatment has become more challenging and costlier. Therefore, this meta-analysis aimed to estimate worldwide resistance rates of genital Mycoplasmas and Ureaplasma to fluoroquinolones (ciprofloxacin, ofloxacin, moxifloxacin, and levofloxacin) agents. We searched the relevant published studies in PubMed, Scopus, and Embase from until 3, March 2022. All statistical analyses were carried out using the statistical package R. The 30 studies included in the analysis were performed in 16 countries. In the metadata, the proportions of ciprofloxacin, ofloxacin, moxifloxacin, and levofloxacin resistance in Mycoplasma and Ureaplasma urogenital isolates were reported 59.8% (95% CI 49.6, 69.1), 31.2% (95% CI 23, 40), 7.3% (95% CI 1, 31), and 5.3% (95% CI 1, 2), respectively. According to the meta-regression, the ciprofloxacin, ofloxacin, moxifloxacin, and levofloxacin rate increased over time. There was a statistically significant difference in the fluoroquinolones resistance rates between different continents/countries (P < 0.05). Based on the results obtained in this systematic review and meta-analysis we recommend the use of the newer group of fluoroquinolones especially levofloxacin as the first choice for the treatment of genital mycoplasmosis, as well as ofloxacin for the treatment of genital infections caused by U. parvum.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ofloxacin hydrochloride, ≥98% (HPLC), powder, fluoroquinolone antibiotic