Merck
  • Exploring the binding mechanism and esterase-like activity of human serum albumin with levofloxacin and its choline based conjugates: A biophysical approach.
Exploring the binding mechanism and esterase-like activity of human serum albumin with levofloxacin and its choline based conjugates: A biophysical approach.
International journal of biological macromolecules (2024-06-10)
Ab Raouf Bhat, Rajan Patel
ABSTRAKT
Human serum albumin (HSA) effectively binds to compounds having different molecular weight and thus facilitates their distribution in the living organisms. Thus, the binding interactions between a potential antibacterial drug (levofloxacin) and synthesized choline based levofloxacinate conjugates with HSA have been explored. The binding efficacy and mechanism were explored by utilizing different spectroscopic techniques; UV-Visible, steady state fluorescence, time resolved fluorescence and esterase-like activity. The interactions between the ligands and protein were electrostatic as well as hydrophobic in nature. The influence of different ligands having different alkyl chain shows quenching of the fluorescence emission of HSA. The spontaneous binding/quenching of HSA with ligands was static in nature, validated by steady state and time resolved fluorescence spectroscopy. Also, the impact of these ligands on the conformation of the native HSA structure was evaluated by using circular dichroism spectroscopy. In combination to the structural change study, the native protein functionality was observed (in terms of 'esterase-like activity') which has been found to be on lower side due to ligand binding. Further, we have performed the reverse study to check the impact of HSA on the fluorescent fluoroquinolone drug. The current study may prove helpful in elucidating the chemico-biological interactions which may prove useful in the pharmaceuticals, pharmacology, and different biochemistry fields.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ofloxacin hydrochloride, ≥98% (HPLC), powder, fluoroquinolone antibiotic