Merck
In vitro and in vivo activity of DL-8280, a new oxazine derivative.
Antimicrobial agents and chemotherapy (1982-10-01)
K Sato, Y Matsuura, M Inoue, T Une, Y Osada, H Ogawa, S Mitsuhashi
ABSTRAKT
DL-8280, 9-fluoro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2,3-dihydro-7H- pyrido-(1,2,3-de)1,4-benzoxazine-6-carboxylic acid, is a new nalidixic acid analog with a broad spectrum of antibacterial activity against gram-negative and gram-positive bacteria, including obligate anaerobes. The activity of DL-8280 against Enterobacteriaceae, Pseudomonas aeruginosa, Haemophilus influenzae, Neisseria gonorrhoeae, and Clostridium perfringens was roughly comparable to that of norfloxacin and far exceeded that of pipemidic acid and nalidixic acid. DL-8280 had greater activity against Staphylococcus spp., Streptococcus spp., Pseudomonas maltophilia, Acinetobacter spp., and Bacteroides fragilis than did norfloxacin, pipemidic acid, and nalidixic acid. Nalidixic acid-resistant Enterobacteriaceae, ampicillin-resistant gonococci, and clindamycin-resistant obligate anaerobes were also susceptible to DL-8280. The activity of DL-8280 was affected very little by inoculum size, and its action was bactericidal at two times the minimal inhibitory concentrations at most. Administered orally to mice experimentally infected with Staphylococcus aureus, Streptococcus pyogenes, Escherichia coli, Proteus mirabilis, Serratia marcescens, or P. aeruginosa, DL-8280 was 2 to 7 times more effective than norfloxacin and 7 to more than 50 times more active than pipemidic acid.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ofloxacin hydrochloride, ≥98% (HPLC), powder, fluoroquinolone antibiotic