- Discovery of a First-in-Class Inhibitor of the Histone Methyltransferase SETD2 Suitable for Preclinical Studies.
Discovery of a First-in-Class Inhibitor of the Histone Methyltransferase SETD2 Suitable for Preclinical Studies.
ACS medicinal chemistry letters (2021-10-22)
John W Lampe, Joshua S Alford, P Ann Boriak-Sjodin, Dorothy Brach, Kat Cosmopoulos, Kenneth W Duncan, Sean T Eckley, Megan A Foley, Darren M Harvey, Vinny Motwani, Michael J Munchhof, Alejandra Raimondi, Thomas V Riera, Cuyue Tang, Michael J Thomenius, Jennifer Totman, Neil A Farrow
PMID34671445
ABSTRAKT
SET domain-containing protein 2 (SETD2), a histone methyltransferase, has been identified as a target of interest in certain hematological malignancies, including multiple myeloma. This account details the discovery of EPZ-719, a novel and potent SETD2 inhibitor with a high selectivity over other histone methyltransferases. A screening campaign of the Epizyme proprietary histone methyltransferase-biased library identified potential leads based on a 2-amidoindole core. Structure-based drug design (SBDD) and drug metabolism/pharmacokinetics (DMPK) optimization resulted in EPZ-719, an attractive tool compound for the interrogation of SETD2 biology that enables in vivo target validation studies.
MATERIAŁY