Merck
GPR34 senses demyelination to promote neuroinflammation and pathologies.
Cellular & molecular immunology (2024-07-20)
Bolong Lin, Yubo Zhou, Zonghui Huang, Ming Ma, Minghui Qi, Zhongjun Jiang, Guoyang Li, Yueli Xu, Jiaxian Yan, Di Wang, Xiaqiong Wang, Wei Jiang, Rongbin Zhou
ABSTRAKT
Sterile neuroinflammation is a major driver of multiple neurological diseases. Myelin debris can act as an inflammatory stimulus to promote inflammation and pathologies, but the mechanism is poorly understood. Here, we showed that lysophosphatidylserine (LysoPS)-GPR34 axis played a critical role in microglia-mediated myelin debris sensing and the subsequent neuroinflammation. Myelin debris-induced microglia activation and proinflammatory cytokine expression relied on its lipid component LysoPS. Both myelin debris and LysoPS promoted microglia activation and the production of proinflammatory cytokines via GPR34 and its downstream PI3K-AKT and ERK signaling. In vivo, reducing the content of LysoPS in myelin or inhibition of GPR34 with genetic or pharmacological approaches reduced neuroinflammation and pathologies in the mouse models of multiple sclerosis and stroke. Thus, our results identify GPR34 as a key receptor to sense demyelination and CNS damage and promote neuroinflammation, and suggest it as a potential therapeutic target for demyelination-associated diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
GPR34 Antagonist, T8848, ≥98% (HPLC)