Merck
Rationally designed high-affinity 2-amino-6-halopurine heat shock protein 90 inhibitors that exhibit potent antitumor activity.
Journal of medicinal chemistry (2007-05-10)
Srinivas R Kasibhatla, Kevin Hong, Marco A Biamonte, David J Busch, Patricia L Karjian, John L Sensintaffar, Adeela Kamal, Rachel E Lough, John Brekken, Karen Lundgren, Roy Grecko, Gregg A Timony, Yingqing Ran, Robert Mansfield, Lawrence C Fritz, Edgar Ulm, Francis J Burrows, Marcus F Boehm
ABSTRAKT
Heat shock protein 90 (Hsp90) is a molecular chaperone protein implicated in stabilizing the conformation and maintaining the function of many cell-signaling proteins. Many oncogenic proteins are more dependent on Hsp90 in maintaining their conformation, stability, and maturation than their normal counterparts. Furthermore, recent data show that Hsp90 exists in an activated form in malignant cells but in a latent inactive form in normal tissues, suggesting that inhibitors selective for the activated form could provide a high therapeutic index. Hence, Hsp90 is emerging as an exciting new target for the treatment of cancer. We now report on a novel series of 2-amino-6-halopurine Hsp90 inhibitors exemplified by 2-amino-6-chloro-9-(4-iodo-3,5-dimethylpyridin-2-ylmethyl)purine (30). These highly potent inhibitors (IC50 of 30 = 0.009 microM in a HER-2 degradation assay) also display excellent antiproliferative activity against various tumor cell lines (IC50 of 30 = 0.03 microM in MCF7 cells). Moreover, this class of inhibitors shows higher affinity for the activated form of Hsp90 compared to our earlier 8-sulfanylpurine Hsp90 inhibitor series. When administered orally to mice, these compounds exhibited potent tumor growth inhibition (>80%) in an N87 xenograft model, similar to that observed with 17-allylamino-17-desmethoxygeldanamycin (17-AAG), which is a compound currently in phase I/II clinical trials.
MATERIAŁY
Numer produktu
Marka
Opis produktu
BIIB021, ≥98% (HPLC), powder, Hsp90 inhibitor