Merck
Advances in the treatment of soft tissue sarcoma: focus on eribulin.
Cancer management and research (2018-02-15)
Panagiotis Koliou, Vasilios Karavasilis, Maria Theochari, Seth M Pollack, Robin L Jones, Khin Thway
ABSTRAKT
Eribulin mesylate is a synthetic derivative of halichondrin B isolated from a marine sponge. Its mechanism of action is through microtubule inhibition, which is different from that of taxanes. Eribulin has been approved for the treatment of metastatic breast cancer and more recently for non-operable or metastatic liposarcoma in patients who have received prior anthracycline chemotherapy. The major side effects of eribulin are bone marrow suppression including neutropenia, leukopenia, anemia, and fatigue/weakness, which can be well managed. In this article, we reviewed evidence from the latest published data on eribulin and its use in the treatment of soft tissue sarcomas. We explored the drug's mechanism of action, pharmacodynamics, pharmacokinetics, and metabolism. Lastly, we reviewed all preclinical studies as well as clinical trials that investigated eribulin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Eribulin, ≥98% (HPLC), powder, microtubule dynamics inhibitor