Merck
  • Effect of processing on proximate composition, anti-nutrient status and amino acid content in three accessions of African locust bean (Parkia biglobosa (jacq.) benth.
Effect of processing on proximate composition, anti-nutrient status and amino acid content in three accessions of African locust bean (Parkia biglobosa (jacq.) benth.
International journal of food sciences and nutrition (2012-07-14)
Ikootobong Sunday Urua, Edak Aniedi Uyoh, Valentine Otang Ntui, Elza Cletus Okpako
ABSTRAKT
Proximate composition, amino acid levels and anti-nutrient factors (polyphenols, phytic acid and oxalate) in the seeds of Parkia biglobosa were determined at three stages: raw, boiled and fermented. The highest anti-nutrient factor present in the raw state was oxalate, while phytic acid was the least. The amino acid of the raw seeds matched favourably to the World Health Organization reference standard. After processing, boiling increased fat, crude fibre and protein, while it reduced moisture, ash and the anti-nutrient content in 64% of the cases examined. Fermentation reduced ash, crude fibre and carbohydrate in all the accessions. It increased the moisture, fat and protein, while reducing the anti-nutrient factors in 78% of the cases. The high levels of protein, fat and amino acids coupled with the low levels of the anti-nutrients in the boiled and fermented seeds make Parkia a good source of nutrients for humans and livestock.