Merck
Study of the size and morphology of aggregates formed by pentaethylene glycol monooctyl ether (C"8EO"5) in n-heptane
Colloids and Surfaces. A, Physicochemical and Engineering Aspects (2012)
Sandoval, et al.