- The discovery of a mevalonate-independent pathway for isoprenoid biosynthesis in bacteria, algae and higher plants.
The discovery of a mevalonate-independent pathway for isoprenoid biosynthesis in bacteria, algae and higher plants.
Natural product reports (1999-12-10)
M Rohmer
PMID10584331