- Iron-substituted polyoxotungstates as inorganic synzymes: evidence for a biomimetic pathway in the catalytic oxygenation of catechols.
Iron-substituted polyoxotungstates as inorganic synzymes: evidence for a biomimetic pathway in the catalytic oxygenation of catechols.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-07-09)
Andrea Sartorel, Mauro Carraro, Gianfranco Scorrano, Bassem S Bassil, Michael H Dickman, Bineta Keita, Louis Nadjo, Ulrich Kortz, Marcella Bonchio
PMID19585647
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protocatechuate 3,4-Dioxygenase from Pseudomonas sp., lyophilized powder, ≥3 units/mg solid