Merck
  • Stimulation of reductive glycerol metabolism by overexpression of an aldehyde dehydrogenase in a recombinant Klebsiella pneumoniae strain defective in the oxidative pathway.
Stimulation of reductive glycerol metabolism by overexpression of an aldehyde dehydrogenase in a recombinant Klebsiella pneumoniae strain defective in the oxidative pathway.
Journal of industrial microbiology & biotechnology (2010-09-24)
Lian Hua Luo, Jeong-Woo Seo, Baek-Rock Oh, Pil-Soo Seo, Sun-Yeon Heo, Won-Kyung Hong, Dae-Hyuk Kim, Chul Ho Kim
ABSTRAKT
Previously, we constructed a glycerol oxidative pathway-deficient mutant strain of Klebsiella pneumoniae by inactivation of glycerol dehydrogenase (dhaD) to eliminate by-product synthesis during production of 1,3-propanediol (1,3-PD) from glycerol. Although by-product formation was successfully blocked in the resultant strain, the yield of 1,3-PD was not enhanced, probably because dhaD disruption resulted in insufficient regeneration of the cofactor NADH essential for the activity of 1,3-PD oxidoreductase (DhaT). To improve cofactor regeneration, in the present study we overexpressed an NAD(+)-dependent aldehyde dehydrogenase in the recombinant strain. To this end, an aldehyde dehydrogenase AldHk homologous to E. coli AldH but with NAD(+)-dependent propionaldehyde dehydrogenase activity was identified in K. pneumoniae. Functional analysis revealed that the substrate specificity of AldHk embraced various aldehydes including propionaldehyde, and that NAD(+) was preferred over NADP(+) as a cofactor. Overexpression of AldHk in the glycerol oxidative pathway-deficient mutant AK/pVOTHk resulted in a 3.6-fold increase (0.57 g l(-1) to 2.07 g l(-1)) in the production of 3-hydroxypropionic acid (3-HP), and a 1.1-fold enhancement (8.43 g l(-1) to 9.65 g l(-1)) of 1,3-PD synthesis, when glycerol was provided as the carbon source, compared to the levels synthesized by the control strain (AK/pVOT). Batch fermentation using AK/pVOTHk showed a significant increase (to 70%, w/w) in conversion of glycerol to the reductive metabolites, 1,3-PD and 3-HP, with no production of by-products except acetate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycerol Dehydrogenase from Cellulomonas sp., lyophilized powder, ≥50 units/mg protein