Merck
Preparation of glutamate analogues by enzymatic transamination.
Methods in molecular biology (Clifton, N.J.) (2011-10-01)
Thierry Gefflaut, Zeinab Assaf, Martine Sancelme
ABSTRAKT
Aminotransferases are key enzymes of the metabolism of proteinogenic amino acids. These ubiquitous biocatalysts show high specific activities and relaxed substrate specificities making them valuable tools for the stereoselective synthesis of unnatural amino acids. We describe here the application of aspartate aminotransferase and branched chain aminotransferase from E. coli for the synthesis of various glutamate analogues, molecules of particular interest regarding the neuroactive properties of glutamic acid.