Merck
Systematic isolation of circulating human peptides: the concept of peptide trapping.
European journal of medical research (1996-02-22)
P Schulz-Knappe, M Raida, M Meyer, E A Quellhorst, W G Forssmann
ABSTRAKT
The structural determination of circulating human peptides is essential to determine their correct posttranslationally processed form. Human hemofiltrate from patients with end stage renal disease is accessible in large quantities and is used as a source for the preparation of circulating peptides. After complete peptide extraction from hemofiltrate, a systematic separation with different chromatographic techniques is achieved. Single peptides are selected according to their mass and chromatographic elution position. Following chromatographic purification, amino acid sequence analysis is performed in combination with data base search. The identification of circulating peptides leads to numerous fragments resulting from cleavage of larger plasma proteins as well as to the discovery of new peptide hormones. The results obtained so far give insight into the degradation of plasma proteins such as fibrinogen, which results in the generation of fragments with biological activity themselves and in the identification of a novel cytokine HCC-1, the first member of beta-defensins in humans, hBD-1, and different peptides not present in any data base.