Merck
Inhibition of HIV-1 infection by the beta-chemokine MDC.
Science (New York, N.Y.) (1997-10-24)
R Pal, A Garzino-Demo, P D Markham, J Burns, M Brown, R C Gallo, A L DeVico
ABSTRAKT
CD8(+) T lymphocytes from individuals infected with human immunodeficiency virus-type 1 (HIV-1) secrete a soluble activity that suppresses infection by HIV-1. A protein associated with this activity was purified from the culture supernatant of an immortalized CD8(+) T cell clone and identified as the beta-chemokine macrophage-derived chemokine (MDC). MDC suppressed infection of CD8(+) cell-depleted peripheral blood mononuclear cells by primary non-syncytium-inducing and syncytium-inducing isolates of HIV-1 and the T cell line-adapted isolate HIV-1IIIB. MDC was expressed in activated, but not resting, peripheral blood mononuclear cells and binds a receptor on activated primary T cells. These observations indicate that beta-chemokines are responsible for a major proportion of HIV-1-specific suppressor activity produced by primary T cells.