Merck
Future needs in exploration of gene-environment interactions.
Journal of hypertension (2012-09-14)
Pavel Hamet
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cathepsin L from human liver, ≥0.5 units/mg protein, solution