Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
D-Ribulose 1,5-Diphosphate Carboxylase from spinach, partially purified powder, 0.01-0.1 unit/mg solid
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)