- Interaction of rifamycin with bacterial RNA polymerase.
Interaction of rifamycin with bacterial RNA polymerase.
Proceedings of the National Academy of Sciences of the United States of America (1968-10-01)
W Wehrli, F Knüsel, K Schmid, M Staehelin
PMID4879400
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals