Merck
[Genetic study for diabetic microvascular complications--recent advances and future perspectives].
Nihon rinsho. Japanese journal of clinical medicine (2012-11-20)
Shiro Maeda
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetyl-CoA Carboxylase 1 human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells