Merck
  • Anthocyanins suppress the cleavable complex formation by irinotecan and diminish its DNA-strand-breaking activity in the colon of Wistar rats.
Anthocyanins suppress the cleavable complex formation by irinotecan and diminish its DNA-strand-breaking activity in the colon of Wistar rats.
Carcinogenesis (2013-01-01)
Melanie Esselen, Stephan W Barth, Swantje Winkler, Simone Baechler, Karlis Briviba, Bernhard Watzl, Susanne Skrbek, Doris Marko
ABSTRAKT
In the present study, the question was addressed whether anthocyanins interfere with the topoisomerase I poison irinotecan in vivo. In vivo complexes of enzyme to DNA bioassay was used to detect irinotecan-induced stabilization of topoisomerase I/DNA complexes and single cell gel electrophoresis to determine DNA-strand-break induction in the colon of male Wistar rats. Furthermore, analysis of anthocyanin concentrations in rat plasma and rat colon was included in the testing, demonstrating that anthocyanins reach the colon and the concentrations do not differ between rats that only received anthocyanins and the anthocyanin/irinotecan group. Blackberry extract was found to significantly reduce irinotecan-mediated topoisomerase I/DNA cleavable complex formation. Overall, anthocyanins did not notably increase cleavable complex formation. However, a significant increase of DNA damage was shown after a single dose of irinotecan as well as the single compounds cyanidin (cy) and cyanidin-3-glucoside (cy-3-g). Furthermore, a significant reduction of irinotecan-induced DNA-strand breaks after a pretreatment with cy, cy-3-g and blackberry extract was observed. Thus, the question arises whether anthocyanin-rich preparations might interfere with chemotherapy or whether, due to low systemic bioavailability, the preparations might provide protective potential in the gastrointestinal tract.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kuromanin chloride, ≥95% (HPLC)
Sigma-Aldrich
Topoisomerase I human, buffered aqueous glycerol solution, ≥2 units/μL