- Evaluation of biotransformed berberine derivatives as anti inflammatory drugs: an in silico study.
Evaluation of biotransformed berberine derivatives as anti inflammatory drugs: an in silico study.
Interdisciplinary sciences, computational life sciences (2013-01-29)
Chandra D Naveen, G K Prasanth, C Sadasivan, M Haridas
PMID23354815
ABSTRAKT
The anti inflammatory potential of the human and microbial biotransformed derivatives of berberine were determined by molecular docking. It was revealed that almost all derivatives formed as a result of biotransformation showed increase in phospholipase A(2) binding affinity compared to berberine. The newly introduced -OH group/groups establish stronger hydrogen bonding interactions and more number of van der Waals contacts with the protein. As phospholipase A(2) is a target of anti inflammatory drugs, it might be concluded that certain biotransformed derivatives of berberine could be better anti inflammatory agents compared to berberine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein