Merck
Highly H2O2-sensitive electrospun quantum dots nanocomposite films for fluorescent biosensor.
Journal of biomedical nanotechnology (2013-05-01)
Longfei Tan, Xiaolong He, Dong Chen, Xiaoli Wu, Hongbo Li, Xiangling Ren, Xianwei Meng, Fangqiong Tang
ABSTRAKT
Bright CdSe quantum dots (QDs)/polycaprolactone (PCL) nanocomposite fluorescent films were fabricated by electronspinning. By using chloroform and N,N-dimethylformamide as electronspinning solvent, the oil-soluble CdSe QDs were uniformly distributed in PCL fibers, and were directly employed as optical probe without any modification processing. The fluorescences of CdSe QDs/PCL nanocomposite films were quickly quenched when the films were contacted with H2O2, solution. In the presence of glucose oxidase (GOD), the fluorescence intensities of these fluorescent films exhibit a liner change with the concentrations of glucose. The H2O2-sensitive electrospun QDs nanocomposite films are highly uniform and repeatable, demonstrating the potential to fabricate stable, sensitive and recyclable fluorescent biosensor for the detection different H2O2-generating oxidases and their substrates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Polycaprolactone, average Mn 45,000
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
Sigma-Aldrich
Polycaprolactone, average Mw ~14,000, average Mn ~10,000 by GPC
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade