Merck
[Sarcosine dehydrogenase deficiency].
Ryoikibetsu shokogun shirizu (1998-05-20)
K Hiraga
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Sarcosine, BioXtra