Merck
Use of a Gluconobacter frateurii mutant to prevent dihydroxyacetone accumulation during glyceric acid production from glycerol.
Bioscience, biotechnology, and biochemistry (2010-11-13)
Hiroshi Habe, Yuko Shimada, Tokuma Fukuoka, Dai Kitamoto, Masayuki Itagaki, Kunihiro Watanabe, Hiroshi Yanagishita, Toshiharu Yakushi, Kazunobu Matsushita, Keiji Sakaki
ABSTRAKT
To prevent dihydroxyacetone (DHA) by-production during glyceric acid (GA) production from glycerol using Gluconobacter frateurii, we used a G. frateurii THD32 mutant, ΔsldA, in which the glycerol dehydrogenase subunit-encoding gene (sldA) was disrupted, but ΔsldA grew much more slowly than the wild type, growth starting after a lag of 3 d under the same culture conditions. The addition of 1% w/v D-sorbitol to the medium improved both the growth and the GA productivity of the mutant, and ΔsldA produced 89.1 g/l GA during 4 d of incubation without DHA accumulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycerol Dehydrogenase from Cellulomonas sp., lyophilized powder, ≥50 units/mg protein