- Enzymatic fluorometric assay for myo-inositol trisphosphate.
Enzymatic fluorometric assay for myo-inositol trisphosphate.
Analytical biochemistry (1987-05-01)
J A Shayman, A R Morrison, O H Lowry
PMID3496810
ABSTRAKT
The determination of myo-inositol trisphosphate by an enzymatic fluorometric assay is presented. The method involves the acid extraction of water-soluble inositol polyphosphates followed by separation by anion-exchange chromatography. Samples are subsequently neutralized by passage over a Dowex Cl- resin and elution with lithium chloride. Samples are then desalted with ethanol. Following dephosphorylation with alkaline phosphatase, free myo-inositol is measured enzymatically via the NAD-dependent oxidation to scyllo-inosose with myo-inositol dehydrogenase. The efficiency of recovery, assay specificity, and an application to the measurement of inositol polyphosphates in hormone-stimulated tissue are discussed.