Merck
Production of non-glycosylated recombinant proteins in Nicotiana benthamiana plants by co-expressing bacterial PNGase F.
Plant biotechnology journal (2012-04-24)
Tarlan Mamedov, Ananya Ghosh, R Mark Jones, Vadim Mett, Christine E Farrance, Konstantin Musiychuk, April Horsey, Vidadi Yusibov
ABSTRAKT
Application of tools of molecular biology and genomics is increasingly leading towards the development of recombinant protein-based biologics. As such, it is leading to an increased diversity of targets that have important health applications and require more flexible approaches for expression because of complex post-translational modifications. For example, Plasmodium parasites may have complex post-translationally modified proteins such as Pfs48/45 that do not carry N-linked glycans (Exp. Parasitol. 1998; 90, 165.) but contain potential N-linked glycosylation sites that can be aberrantly glycosylated during expression in mammalian and plant systems. Therefore, it is important to develop strategies for producing non-glycosylated forms of these targets to preserve biological activity and native conformation. In this study, we are describing in vivo deglycosylation of recombinant N-glycosylated proteins as a result of their transient co-expression with bacterial PNGase F (Peptide: N-glycosidase F). In addition, we show that the recognition of an in vivo deglycosylated plant-produced malaria vaccine candidate, Pfs48F1, by monoclonal antibodies I, III and V raised against various epitopes (I, III and V) of native Pfs48/45 of Plasmodium falciparum, was significantly stronger compared to that of the glycosylated form of plant-produced Pfs48F1. To our knowledge, neither in vivo enzymatic protein deglycosylation has been previously achieved in any eukaryotic system, including plants, nor has bacterial PNGase F been expressed in the plant system. Thus, here, we report for the first time the expression in plants of an active bacterial enzyme PNGase F and the production of recombinant proteins of interest in a non-glycosylated form.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli