Merck
5-HT2C receptor mediation of unconditioned escape behaviour in the unstable elevated exposed plus maze.
Psychopharmacology (2002-10-31)
Nicholas Jones, Mark S Duxon, Sheila M King
ABSTRAKT
m-Chlorophenylpiperazine (mCPP) induces panic in humans and dose dependently increases unconditioned escape behaviour in a novel pre-clinical model of extreme anxiety in rats, the unstable elevated exposed plus maze (UEEPM). Numerous studies indicate that the anxiogenic effects of mCPP may be mediated by its action at the 5-HT2C receptor. This study aimed to examine the involvement of the 5-HT2C receptor in the unconditioned fear responses observed in the UEEPM (after an acute dose of mCPP) by pre-treatment with the selective 5-HT2C receptor antagonist SB-242084. Male Hooded Lister rats received a single dose of SB-242084 (0.1-1.0 mg/kg IP) or vehicle 40 min pre-test followed by a single dose of mCPP (1.0 mg/kg IP) or saline 30 min before being exposed to the UEEPM for a period of 5 min. Subjects' behaviour was analysed to determine the effects of SB-242084 on mCPP-induced increases in escape behaviour. mCPP alone increased animals' propensity to escape from the UEEPM despite producing marked decreases in locomotor/exploratory behaviour. SB-242084 dose dependently inhibited the increases in escape and hypolocomotor effects induced by mCPP. These results suggest that the escape-related behaviours exhibited by animals in the UEEPM are mediated, at least in part, by activation of the 5-HT2C receptor subtype.