Merck
Molecular characterization of cytosolic phospholipase A2-beta.
The Journal of biological chemistry (1999-06-08)
C Song, X J Chang, K M Bean, M S Proia, J L Knopf, R W Kriz
ABSTRAKT
We have isolated a cDNA encoding a 1012-amino acid polypeptide cPLA2-beta, that has significant homology with cPLA2-alpha in both the calcium-dependent lipid binding domain as well as in the catalytic domain. Transient expression of cPLA2-beta cDNA in COS cells results in an increase in calcium-dependent phospholipase A1 (PLA1) and PLA2 activities compared with vector-transfected cells. cPLA2-beta is markedly less selective for cleavage at sn-2 as compared with cPLA2-alpha and cPLA2-gamma. Northern analysis reveals a cPLA2-beta transcript of 8 kilobase pairs that is expressed in all the human tissues examined. With the identification of cPLA2-beta, the newly defined cPLA2 family now comprises three members that may have dramatically different mechanisms for regulation of expression and enzymatic activation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein